missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-cyclopenten-1-one, 97%, Thermo Scientific Chemicals

Catalog Number p-161763
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1120-73-6
C6H8O
96.13
MFCD00012275
ZSBWUNDRDHVNJL-UHFFFAOYSA-N
2-methylcyclopent-2-enone, 2-methyl-2-cyclopenten-1-one, 2-cyclopenten-1-one, 2-methyl, 2-methyl-2-cyclopentenone, cyclopentenone, 2-methyl, cyclopentenone, methyl, methyl-cyclopentenone, acmc-20af73, 2-cyclopenten-1-one, methyl, 2-cyclopenten-1-one,2-methyl
14266
2-methylcyclopent-2-en-1-one
CC1=CCCC1=O
This item is not returnable. View return policy
1120-73-6
C6H8O
96.13
MFCD00012275
ZSBWUNDRDHVNJL-UHFFFAOYSA-N
2-methylcyclopent-2-enone, 2-methyl-2-cyclopenten-1-one, 2-cyclopenten-1-one, 2-methyl, 2-methyl-2-cyclopentenone, cyclopentenone, 2-methyl, cyclopentenone, methyl, methyl-cyclopentenone, acmc-20af73, 2-cyclopenten-1-one, methyl, 2-cyclopenten-1-one,2-methyl
14266
2-methylcyclopent-2-en-1-one
CC1=CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9790g/mL
Boiling Point 158.0°C to 161.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C5H5(0)
Refractive Index 1.4780 to 1.4800
Quantity 1 g
Beilstein 07, 52
Specific Gravity 0.979
Formula Weight 96.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-cyclopenten-1-one, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-2-cyclopenten-1-one
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'