missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1H-imidazole-4-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Formula Weight | 110.12 |
Chemical Name or Material | 2-Methyl-1H-imidazole-4-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'