2-Methyl-1-propanol, 99+%, pure, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 398950010

1LT 2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 78-83-1 Weight: 0.85000kg Transport: UN number : 1212 Chem class : 1 Pack group : III


Packaging Glass Bottle
Quantity 1L

Product Code. 13141944

Quantity Price
1 £ 60.94 / 1 L
EU Stock 10
Add to basket

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Methyl-1-propanol
Name Note Extra Dry
CAS 78-83-1
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Pure
Assay Percent Range 99+%
Molecular Formula C4 H10 O
Linear Formula (CH3)2CHCH2OH
Formula Weight 74.12
MDL Number MFCD00004740
Synonym Isobutyl alcohol

Specification

Formula Weight 74.12
Quantity 1L
Chemical Name or Material 2-Methyl-1-propanol
Packaging Glass Bottle
Name Note Extra Dry
Purity Grade Pure
Assay Percent Range 99+%
Molecular Formula C4 H10 O
Linear Formula (CH3)2CHCH2OH
MDL Number MFCD00004740
CAS 78-83-1
Beilstein 01, 373
Merck Index 15, 5176
Synonym Isobutyl alcohol
CAS Min % 99.0
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Density 0.8000g/mL
Documentation Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.