missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-buten-3-yne, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Methyl-1-buten-3-yne is a reagent used in analytical studies to determine the aroma compounds present in various things by using pyrolysis gas chromatography mass spectrometry techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-1-buten-3-yne is a reagent used in analytical studies to determine the aroma compounds present in various things by using pyrolysis gas chromatography mass spectrometry techniques.
Solubility
Not miscible in water.
Notes
Store at 4°C. Incompatible with heavy metal powders, heat. Protect from heat and direct sunlight. Store away from oxidizing agents.
2-Methyl-1-buten-3-yne is a reagent used in analytical studies to determine the aroma compounds present in various things by using pyrolysis gas chromatography mass spectrometry techniques.
Solubility
Not miscible in water.
Notes
Store at 4°C. Incompatible with heavy metal powders, heat. Protect from heat and direct sunlight. Store away from oxidizing agents.
Specifications
Melting Point | -113°C |
Density | 0.694 |
Boiling Point | 32°C to 33°C |
Flash Point | −45°C (−49°F) |
Odor | Pungent |
Refractive Index | 1.418 |
Quantity | 5 g |
UN Number | UN3295 |
Beilstein | 969175 |
Solubility Information | Not miscible in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'