missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals

Catalog Number 10626534
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
58-27-5
C11H8O2
172.18
MFCD00001681
MJVAVZPDRWSRRC-UHFFFAOYSA-N
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
4055
CHEBI:28869
2-methylnaphthalene-1,4-dione
CC1=CC(=O)C2=CC=CC=C2C1=O
This item is not returnable. View return policy
58-27-5
C11H8O2
172.18
MFCD00001681
MJVAVZPDRWSRRC-UHFFFAOYSA-N
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
4055
CHEBI:28869
2-methylnaphthalene-1,4-dione
CC1=CC(=O)C2=CC=CC=C2C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 105.0°C to 108.0°C
Color Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 07, II, 656
Merck Index 15, 5899
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol (1g/60 mL),benzene (1g/10 mL,soluble in vegetable oils (1g/50 mL),moderately soluble in chloroform and carbon tetrac
Formula Weight 172.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Methyl-1, 4-naphthoquinone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-1,4-naphthoquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'