missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-(4-methylpiperidino)-2-propanamine, 97%, Maybridge

Brand:  Maybridge MO01186DE

Code : 31

Additional Details : CAS Number : 690632-11-2 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 2b Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 10515675

  • £329.57 / 5g


Estimated Shipment date
from Supplier 23-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-1-(4-methylpiperidino)-2-propanamine
97.0
97%
170.3
2-Methyl-1-(4-methylpiperidino)-2-propanamine (IUPAC)
690632-11-2
97%
C10H22N2
MFCD05022424

Specifications

2-Methyl-1-(4-methylpiperidino)-2-propanamine
97.0
97%
170.3
2-Methyl-1-(4-methylpiperidino)-2-propanamine (IUPAC)
690632-11-2
97%
C10H22N2
MFCD05022424
5g
Videos
SDS