missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2698572
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
35272-15-2
C5H5NO2S
143.16
MFCD03407332
ZHDRDZMTEOIWSX-UHFFFAOYSA-N
2-methylthiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylic acid, 4-thiazolecarboxylic acid, 2-methyl, 2-methyl-thiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylicacid, 2-methyl-thiazole4-carboxylic acid, 4-thiazolecarboxylicacid, 2-methyl, 2-methyl-1,3-thiazole-4-carboxylicacid
284728
2-methyl-1,3-thiazole-4-carboxylic acid
CC1=NC(=CS1)C(=O)O
This item is not returnable. View return policy
35272-15-2
C5H5NO2S
143.16
MFCD03407332
ZHDRDZMTEOIWSX-UHFFFAOYSA-N
2-methylthiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylic acid, 4-thiazolecarboxylic acid, 2-methyl, 2-methyl-thiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylicacid, 2-methyl-thiazole4-carboxylic acid, 4-thiazolecarboxylicacid, 2-methyl, 2-methyl-1,3-thiazole-4-carboxylicacid
284728
2-methyl-1,3-thiazole-4-carboxylic acid
CC1=NC(=CS1)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'