missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carboximidamide hydrochloride, 97%, Thermo Scientific™

Catalog Number p-2698569
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
18876-82-9
C5H8ClN3S
177.65
MFCD00178765
YYLYRAUCGUIGIE-UHFFFAOYSA-N
2-methyl-1,3-thiazole-4-carboximidamide hydrochloride, 2-methylthiazole-4-carboxamidine hydrochloride, 2-methyl-thiazole-4-carboxamidine hydrochloride, 2-methylthiazole-4-carboximidamide hydrochloride, 2-methyl-1,3-thiazole-4-carboxamidine, chloride, 4-thiazolecarboximidamide,2-methyl-,hydrochloride 1:1
2799402
2-methyl-1,3-thiazole-4-carboximidamide;hydrochloride
CC1=NC(=CS1)C(=N)N.Cl
This item is not returnable. View return policy
18876-82-9
C5H8ClN3S
177.65
MFCD00178765
YYLYRAUCGUIGIE-UHFFFAOYSA-N
2-methyl-1,3-thiazole-4-carboximidamide hydrochloride, 2-methylthiazole-4-carboxamidine hydrochloride, 2-methyl-thiazole-4-carboxamidine hydrochloride, 2-methylthiazole-4-carboximidamide hydrochloride, 2-methyl-1,3-thiazole-4-carboxamidine, chloride, 4-thiazolecarboximidamide,2-methyl-,hydrochloride 1:1
2799402
2-methyl-1,3-thiazole-4-carboximidamide;hydrochloride
CC1=NC(=CS1)C(=N)N.Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 177.66
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carboximidamide hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'