missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carbothioamide, 97%, Maybridge™

Brand:  Maybridge SPB02802DE

Code : 31

Additional Details : CAS Number : 174223-29-1 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Packaging Amber Glass Bottle
Quantity 5g

 View more versions of this product

Product Code. 10275424

  • £239.00 / 5g

Estimated Shipment: 20-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

174223-29-1
158.237
DIOPPULTIGEDCB-UHFFFAOYSA-N
2744365
CC1=NC(=CS1)C(=S)N
C5H6N2S2
MFCD00052167
2-methylthiazole-4-carbothioamide, amino 2-methyl 1,3-thiazol-4-yl methane-1-thione, maybridge4_004334, 2-methylthiazole-4-thiocarboxamide, 4-thiazolecarbothioamide,2-methyl, 4-thiazolecarbothioamide, 2-methyl, 2-methylthiazole-4-carbothioic acid amide, 4-thiazolecarbothioamide, 2-methyl-9ci
2-methyl-1,3-thiazole-4-carbothioamide

Specifications

2-Methyl-1, 3-thiazole-4-carbothioamide
5g
97.0
C5H6N2S2
2-methylthiazole-4-carbothioamide, amino 2-methyl 1,3-thiazol-4-yl methane-1-thione, maybridge4_004334, 2-methylthiazole-4-thiocarboxamide, 4-thiazolecarbothioamide,2-methyl, 4-thiazolecarbothioamide, 2-methyl, 2-methylthiazole-4-carbothioic acid amide, 4-thiazolecarbothioamide, 2-methyl-9ci
CC1=NC(=CS1)C(=S)N
158.237
158.25
Amber Glass Bottle
174223-29-1
97%
MFCD00052167
DIOPPULTIGEDCB-UHFFFAOYSA-N
2-methyl-1,3-thiazole-4-carbothioamide
2744365
97%
Videos
SDS Documents
Certificates