missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,3-dithiane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Colorless to Brown |
Density | 1.1200g/mL |
Boiling Point | 66°C (5.0 Torr) |
Flash Point | 82°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C5H10S2 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'