missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-benzothiazole-6-sulfonyl chloride, 95%, Maybridge

Brand:  Maybridge CC54703DA

Code : 31

Additional Details : CAS Number : 21431-13-0 Weight : 0.05100kg Transport : UN number : 3261 Chem class : 1a Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10786514

  • £100.00 / 1g


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Specifications

Chemical Identifiers

2-Methyl-1, 3-benzothiazole-6-sulfonyl chloride
95%
247.73
95%
C8H6ClNO2S2

Specifications

2-Methyl-1, 3-benzothiazole-6-sulfonyl chloride
21431-13-0
95%
C8H6ClNO2S2
1g
Amber Glass Bottle
95%
247.73
Videos
SDS