missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-benzothiazol-6-ylamine, 97%, Thermo Scientific™

Catalog Number p-4510719
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2941-62-0
C8H8N2S
164.226
HFUJOSYKJMNSFQ-UHFFFAOYSA-N
2-methylbenzo d thiazol-6-amine, 6-amino-2-methylbenzothiazole, 2-methyl-benzothiazol-6-ylamine, 6-benzothiazolamine, 2-methyl, 2-methyl-1,3-benzothiazol-6-ylamine, 6-amino-2-methyl-benzothiazole, 2-methylbenzothiazol-6-amine, 2-methylbenzothiazole-6-ylamine, pubchem20725, acmc-1cctf
762831
2-methyl-1,3-benzothiazol-6-amine
CC1=NC2=C(S1)C=C(C=C2)N
This item is not returnable. View return policy
2941-62-0
C8H8N2S
164.226
HFUJOSYKJMNSFQ-UHFFFAOYSA-N
2-methylbenzo d thiazol-6-amine, 6-amino-2-methylbenzothiazole, 2-methyl-benzothiazol-6-ylamine, 6-benzothiazolamine, 2-methyl, 2-methyl-1,3-benzothiazol-6-ylamine, 6-amino-2-methyl-benzothiazole, 2-methylbenzothiazol-6-amine, 2-methylbenzothiazole-6-ylamine, pubchem20725, acmc-1cctf
762831
2-methyl-1,3-benzothiazol-6-amine
CC1=NC2=C(S1)C=C(C=C2)N
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 164.23
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-benzothiazol-6-ylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'