missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,3-benzothiazol-6-ylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 164.23 |
Percent Purity | 97% |
Chemical Name or Material | 2-Methyl-1, 3-benzothiazol-6-ylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'