missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyrazine, 95%, Thermo Scientific™

Catalog Number 11967041
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 3149-28-8
Density 1.1300g/mL
Boiling Point 60°C to 61°C (29.0 mmHg)
Flash Point 45°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C5H6N2O
Quantity 5 g
Specific Gravity 1.13
Synonym pyrazine, methoxy, methoxypyrazine, 2-methoxy-1,4-diazine, pyrazine, 2-methoxy, 2-methoxy-pyrazine, unii-ryd35t7f4t, 2-methoxy pyrazine, fema no. 3302, ryd35t7f4t, methoxy-pyrazine
Solubility Information Solubility in water: in water:(20°C) slightly soluble
InChI Key WKSXRWSOSLGSTN-UHFFFAOYSA-N
SMILES COC1=NC=CN=C1
IUPAC Name 2-methoxypyrazine
Molecular Weight (g/mol) 110.12
PubChem CID 18467
Formula Weight 110.12
Percent Purity 95%
Chemical Name or Material 2-Methoxypyrazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxypyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'