missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl isothiocyanate, 99%, ACROS Organics™

Brand:  Acros Organics 300220250

Code : 29

Additional Details : CAS Number : 3288-04-8 Weight : 0.07500kg Transport : UN number : 2810 Chem class : 1 Pack group : II

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10792871

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Methoxyphenyl isothiocyanate
3288-04-8
100.0
99%
165.22
o-Anisyl isothiocyanate
99%
98.5
≥98.5%
C8H7NOS
MFCD00011675

Specifications

2-Methoxyphenyl isothiocyanate
3288-04-8
100.0
Authentic
Liquid
99%
C8H7NOS
MFCD00011675
25g
99%
98.5
Glass bottle
1.6433 to 1.6453 (20°C, 589nm)
≥98.5%
1.1900g/mL
165.22
Clear light yellow
o-Anisyl isothiocyanate
Videos
SDS