missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl isothiocyanate, 99%, ACROS Organics™

Brand:  Acros Organics 300220050

Code : 29

Additional Details : CAS Number : 3288-04-8 Weight : 0.05500kg Transport : UN number : 2810 Chem class : 1 Pack group : II

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10450531

  • £29.30 / 5g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

2-Methoxyphenyl isothiocyanate
3288-04-8
100.0
99%
165.22
o-Anisyl isothiocyanate
99%
98.5
99%
C8H7NOS
MFCD00011675

Specifications

2-Methoxyphenyl isothiocyanate
3288-04-8
100.0
Authentic
1.19
99%
1.1900g/mL
165.22
264.0° to 266.0°C
5g
o-Anisyl isothiocyanate
99%
98.5
Glass bottle
1.6433 to 1.6453
Liquid
99%
C8H7NOS
MFCD00011675
Clear light yellow
>110°C
Videos
SDS