missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynicotinic acid, 97%, Thermo Scientific™

Catalog Number 10642895
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
16498-81-0
C7H6NO3
152.13
MFCD00085110
FTEZJSXSARPZHJ-UHFFFAOYSA-M
2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh
708093
COC1=NC=CC=C1C([O-])=O
This item is not returnable. View return policy
16498-81-0
C7H6NO3
152.13
MFCD00085110
FTEZJSXSARPZHJ-UHFFFAOYSA-M
2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh
708093
COC1=NC=CC=C1C([O-])=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name 2-methoxypyridine-3-carboxylic acid
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Methoxynicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'