missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog Number p-7058379
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
3121-61-7
C6H10O3
130.14
MFCD00048149,MFCD00803685
HFCUBKYHMMPGBY-UHFFFAOYSA-N
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
18392
2-methoxyethyl prop-2-enoate
COCCOC(=O)C=C
This item is not returnable. View return policy
3121-61-7
C6H10O3
130.14
MFCD00048149,MFCD00803685
HFCUBKYHMMPGBY-UHFFFAOYSA-N
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
18392
2-methoxyethyl prop-2-enoate
COCCOC(=O)C=C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point <-30°C
Density 1.008
Boiling Point 162°C
Flash Point 56°C (133°F)
Odor Ester-like
Linear Formula H2C=CHCO2(CH2)2OCH3
Refractive Index 1.428
Quantity 50 g
UN Number UN2929
Beilstein 1754333
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material 2-Methoxyethyl acrylate, stabilized with ≈50 to 100ppm 4-methoxyphenol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'