missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyethanol, Extra Pure, SLR, Fisher Chemical™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Spécification
Melting Point | -85°C |
Color | Colorless |
Boiling Point | 124°C |
Packaging | Amber glass bottle |
Quantity | 1 L |
Formula Weight | 76.09g/mol |
Vapor Pressure | 9.5mmHg at 25°C |
Physical Form | Liquid |
Sécurité et traitement
Identifiant produit
- 2-Methoxyethanol
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
- H370-Causes damage to organs.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'