missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

£144.42

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15568814
View Documents
JT Baker
P784-07
500mL Narrow Mouth Amber Glass Bottle £144.42
500mL View alternative products
Save Up to 
»
Estimated Shipment
16-12-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

109-86-4
76.095
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
CHEBI:46790
COCCO
C3H8O2
XNWFRZJHXBZDAG-UHFFFAOYSA-N
8019
2-methoxyethanol

Specifications

2-Methoxyethanol
Narrow Mouth Amber Glass Bottle
500mL
C3H8O2
XNWFRZJHXBZDAG-UHFFFAOYSA-N
2-methoxyethanol
8019
76.1
39.00°C
1L = 0.96kg
109-86-4
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
COCCO
76.095
CHEBI:46790
BAKER ANALYZED™ ACS Reagent