missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, ACS, 99.3+%, Thermo Scientific Chemicals

Catalog Number p-7082626
Click to view available options
Quantity:
4 L
4 x 4 L
This item is not returnable. View return policy
109-86-4
C3H8O2
76.095
MFCD00002867
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
8019
CHEBI:46790
2-methoxyethanol
COCCO
This item is not returnable. View return policy
109-86-4
C3H8O2
76.095
MFCD00002867
XNWFRZJHXBZDAG-UHFFFAOYSA-N
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
8019
CHEBI:46790
2-methoxyethanol
COCCO

2-Methoxy ethanol is used as solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -85°C
Color Colorless
Density 0.965
pH 4 to 7
Boiling Point 124°C to 125°C
Flash Point 46°C (115°F)
Odor Characteristic
Linear Formula CH3OCH2CH2OH
Refractive Index 1.4025
Quantity 4 L
UN Number UN1188
Beilstein 1731074
Merck Index 14,6038
Solubility Information Miscible with water,alcohol,glycerol,ether,acetone,DMFMiscible with water,alcohol,ether,acetone,hydrocarbons,ketones,glycols and dimethylformamide.
Formula Weight 76.1
Percent Purity ≥99.3%
Grade ACS
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'