missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzenethiol, 97%, ACROS Organics™

Brand:  Acros Organics 179980050

Code : 29

Additional Details : CAS Number : 7217-59-6 Weight : 0.06000kg


Quantity 5mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10215470

  • £24.40 / 5mL


Estimated Shipment
13-03-2018
Add to basket


Specifications

Chemical Identifiers

2-Methoxybenzenethiol
7217-59-6
100.0
97%
CH3OC6H4SH
MFCD00004834
97%
96.0
97%
C7H8OS
140.2
2-Methoxythiophenol

Specifications

2-Methoxybenzenethiol
95°C
Clear yellow
97%
96.0
Authentic
97%
C7H8OS
140.2
Glass bottle
2-Methoxythiophenol
1.1400g/mL
225.0° to 228.0°C
5mL
7217-59-6
100.0
Liquid
97%
CH3OC6H4SH
MFCD00004834
1.5905 to 1.5925
1.14
Videos
SDS