missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methoxy-N-n-propylbenzamide, 97%

Brand:  Alfa Aesar™ H59834.03

Code : NEW

Additional Details : CAS Number : 550300-22-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446258

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Methoxy-N-n-propylbenzamide
550300-22-6
193.25
MFCD01611811

Specifications

2-Methoxy-N-n-propylbenzamide
1g
Liquid
193.25
Yellow
550300-22-6
97%
MFCD01611811
Videos
SDS