missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methoxy-N-n-pentylbenzamide, 97%

Brand:  Alfa Aesar™ H59093.03

Code : NEW

Additional Details : CAS Number : 544699-31-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495288

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Methoxy-N-n-pentylbenzamide
544699-31-2
221.3
MFCD03383761

Specifications

2-Methoxy-N-n-pentylbenzamide
1g
Liquid
221.3
Yellow
544699-31-2
97%
MFCD03383761
Videos
SDS