missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylphenyl isocyanate, 98%, Thermo Scientific™

Catalog Number 11925701
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
59741-04-7
C9H9NO2
163.18
IDOHLSFNBKNRRJ-UHFFFAOYSA-N
2-methoxy-5-methylphenyl isocyanate, 2-methoxy-5-methylphenylisocyanate, 2-isocyanato-1-methoxy-4-methyl-benzene, benzene, 2-isocyanato-1-methoxy-4-methyl, acmc-20anrw, 2-methoxy-5-methylbenzenisocyanate, 2-methoxy-5-methylpheny lisocyanate, 2-isocyanato-1-methyloxy-4-methylbenzene, benzene, 2-isocyanato-1-methoxy-4-methyl-9ci
4141520
2-isocyanato-1-methoxy-4-methylbenzene
CC1=CC(=C(C=C1)OC)N=C=O
This item is not returnable. View return policy
59741-04-7
C9H9NO2
163.18
IDOHLSFNBKNRRJ-UHFFFAOYSA-N
2-methoxy-5-methylphenyl isocyanate, 2-methoxy-5-methylphenylisocyanate, 2-isocyanato-1-methoxy-4-methyl-benzene, benzene, 2-isocyanato-1-methoxy-4-methyl, acmc-20anrw, 2-methoxy-5-methylbenzenisocyanate, 2-methoxy-5-methylpheny lisocyanate, 2-isocyanato-1-methyloxy-4-methylbenzene, benzene, 2-isocyanato-1-methoxy-4-methyl-9ci
4141520
2-isocyanato-1-methoxy-4-methylbenzene
CC1=CC(=C(C=C1)OC)N=C=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 25.0°C to 26.0°C
Color Colorless
Density 1.1120g/mL
Boiling Point 240.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.112
Formula Weight 163.18
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Methoxy-5-methylphenyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxy-5-methylphenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'