missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-5-methylbenzeneboronic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Reactant for coupling reactions, Preparation of selective quinazolinyl-phenol inhibitors of CHK1 as potential antitumors and radioprotectants, Rh-catalyzed asymmetric arylation of aldehydes, Stereoselective hydroarylation reactions, Suzuki-Miyaura cross-coupling.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Reactant for coupling reactions, Preparation of selective quinazolinyl-phenol inhibitors of CHK1 as potential antitumors and radioprotectants, Rh-catalyzed asymmetric arylation of aldehydes, Stereoselective hydroarylation reactions, Suzuki-Miyaura cross-coupling.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 80°C to 84°C |
Quantity | 1 g |
Solubility Information | Soluble in methanol. |
Formula Weight | 165.99 |
Percent Purity | 95% |
Chemical Name or Material | 2-Methoxy-5-methylbenzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'