missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals

Catalog Number 10594281
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
120-71-8
C8H11NO
137.18
MFCD00007815
WXWCDTXEKCVRRO-UHFFFAOYSA-N
p-cresidine, cresidine, 5-methyl-o-anisidine, para-cresidine, benzenamine, 2-methoxy-5-methyl, krezidin, 2-amino-4-methylanisole, 3-amino-4-methoxytoluene, 4-methoxy-m-toluidine, 6-methoxy-m-toluidine
8445
CHEBI:82307
2-methoxy-5-methylaniline
CC1=CC(=C(C=C1)OC)N
This item is not returnable. View return policy
120-71-8
C8H11NO
137.18
MFCD00007815
WXWCDTXEKCVRRO-UHFFFAOYSA-N
p-cresidine, cresidine, 5-methyl-o-anisidine, para-cresidine, benzenamine, 2-methoxy-5-methyl, krezidin, 2-amino-4-methylanisole, 3-amino-4-methoxytoluene, 4-methoxy-m-toluidine, 6-methoxy-m-toluidine
8445
CHEBI:82307
2-methoxy-5-methylaniline
CC1=CC(=C(C=C1)OC)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 49.0°C to 55.0°C
Color Brown to Black
Density 111g/mL
Boiling Point 235.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(CH3)NH2
Quantity 5 g
Beilstein 13, II, 338
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most common organic solvents
Formula Weight 137.18
Percent Purity 95%
Physical Form Crystalline Chunks
Chemical Name or Material 2-Methoxy-5-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'