missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-vinylphenol, 97%, stab. with 0.01% BHT, Thermo Scientific™

Catalog Number p-7021028
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7786-61-0
C9H10O2
150.177
MFCD00015437
YOMSJEATGXXYPX-UHFFFAOYSA-N
2-methoxy-4-vinylphenol, 4-vinylguaiacol, 4-hydroxy-3-methoxystyrene, p-vinylguaiacol, phenol, 4-ethenyl-2-methoxy, 2-methoxy-4-vinyl-phenol, para-vinylguaiacol, guaiacol, 4-vinyl, phenol, 2-methoxy-4-vinyl, o-methoxy-p-vinylphenol
332
CHEBI:42438
4-ethenyl-2-methoxyphenol
COC1=C(C=CC(=C1)C=C)O
This item is not returnable. View return policy
7786-61-0
C9H10O2
150.177
MFCD00015437
YOMSJEATGXXYPX-UHFFFAOYSA-N
2-methoxy-4-vinylphenol, 4-vinylguaiacol, 4-hydroxy-3-methoxystyrene, p-vinylguaiacol, phenol, 4-ethenyl-2-methoxy, 2-methoxy-4-vinyl-phenol, para-vinylguaiacol, guaiacol, 4-vinyl, phenol, 2-methoxy-4-vinyl, o-methoxy-p-vinylphenol
332
CHEBI:42438
4-ethenyl-2-methoxyphenol
COC1=C(C=CC(=C1)C=C)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Methoxy-4-vinylphenol is used as a flavoring agent. It is also used as an aromatic substance resembling buckwheat. Further, it is used for chemical signaling in insects such as Red palm weevil. It is also utilized in imitation vanilla, coffee and cocoa.

Solubility
Miscible with water.

Notes
Air, light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 25°C to 29°C
Density 1.11
Boiling Point 108°C to 110°C (3 mmHg)
Flash Point >100°C (212°F)
Odor Pleasant
Refractive Index 1.5818
Quantity 1 g
Beilstein 2044521
Solubility Information Miscible with water.
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-vinylphenol, stabilized with 0.01% BHT
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'