missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™

Catalog Number p-7030716
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
1017779-22-4
C10H9F3O3
234.17
MFCD09832353
GTVKLECZJRZMQO-UHFFFAOYSA-N
2-methoxy-4-trifluoromethyl phenylacetic acid, 2-2-methoxy-4-trifluoromethyl phenyl acetic acid, 2-methoxy-4-trifluoromethyl phenyl acetic acid, 2-methoxy-4-trifluoromethyl phenylacetic acid, jrd
46737670
2-[2-methoxy-4-(trifluoromethyl)phenyl]acetic acid
COC1=C(CC(O)=O)C=CC(=C1)C(F)(F)F
This item is not returnable. View return policy
1017779-22-4
C10H9F3O3
234.17
MFCD09832353
GTVKLECZJRZMQO-UHFFFAOYSA-N
2-methoxy-4-trifluoromethyl phenylacetic acid, 2-2-methoxy-4-trifluoromethyl phenyl acetic acid, 2-methoxy-4-trifluoromethyl phenyl acetic acid, 2-methoxy-4-trifluoromethyl phenylacetic acid, jrd
46737670
2-[2-methoxy-4-(trifluoromethyl)phenyl]acetic acid
COC1=C(CC(O)=O)C=CC(=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 99.5°C to 102°C
Quantity 250 mg
Formula Weight 234.18
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-(trifluoromethyl)phenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'