missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.
Notes
Moisture sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.
Notes
Moisture sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
Specifications
Density | 0.966 |
Boiling Point | 120°C (228 mmHg) |
Flash Point | 46°C (114°F) |
Refractive Index | 1.41 |
Quantity | 5 g |
UN Number | UN1993 |
Beilstein | 2827758 |
Sensitivity | Moisture sensitive |
Formula Weight | 158.01 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'