missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine-N-oxide, sodium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog Number 10013310
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
207511-13-5, 3811-73-2
C5H4NNaOS
149.14
MFCD00151244
WNGMMIYXPIAYOB-UHFFFAOYSA-M
sodium 2-mercaptopyridine n-oxide hydrate
43835086
sodium;1-oxidopyridin-1-ium-2-thiolate;hydrate
[Na+].[O-][N+]1=CC=CC=C1[S-]
This item is not returnable. View return policy
207511-13-5, 3811-73-2
C5H4NNaOS
149.14
MFCD00151244
WNGMMIYXPIAYOB-UHFFFAOYSA-M
sodium 2-mercaptopyridine n-oxide hydrate
43835086
sodium;1-oxidopyridin-1-ium-2-thiolate;hydrate
[Na+].[O-][N+]1=CC=CC=C1[S-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >260°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Merck Index 10, 7892
Solubility Information Solubility in water: soluble
Formula Weight 149.15
Percent Purity 98%
Physical Form Granular Crystalline Powder
Chemical Name or Material 2-Mercaptopyridine-N-oxide,sodium salt hydrate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'