Learn More
2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 257740025
195.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 29
Additional Details : CAS Number : 3811-73-2 Weight : 3.05000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Mercaptopyridine-N-oxide, sodium salt | |
3811-73-2,7732-18-5 | |
60.0 | |
Amber to Yellow | |
8.5 to 10.5 (10% soln. at 20°C) | |
Glass bottle | |
MFCD01941547 | |
1.22 | |
sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van | |
XNRNJIIJLOFJEK-UHFFFAOYSA-N | |
sodium;1-oxidopyridine-2-thione | |
19658 | |
40 to 42% |
40 w/w% Aqueous Solution | |
58.0 | |
-30.0°C | |
1.2200g/mL | |
109.0°C | |
C5H4NNaOS | |
2.5 L | |
10, 7892 | |
Solubility in water: soluble | |
C1=CC(=S)N(C=C1)[O-].[Na+] | |
149.15 | |
149.15 | |
Solution |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH070-Toxic by eye contact.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only