missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine-N-oxide, 99%, ACROS Organics™

Brand:  Acros Organics 182670050

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 1121-31-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10512401

  • £33.70 / 5g


EU Stock 1
Add to basket


Specifications Description & Specifications

Chemical Identifiers

2-Mercaptopyridine-N-oxide
98.5
≥98.5%
C5H5NOS
MFCD00006196
1121-31-9
100.0
99%
127.16
1-Hydroxy-2(1H)-pyridinethione

Specifications

2-Mercaptopyridine-N-oxide
98.5
Authentic
Powder, Crystals, and/or Chunks
99%
127.16
21, V,7, 150
14, 7994
Off-white to beige or yellow to yellow-greenish or grayish-green
5g
1121-31-9
100.0
Glass bottle
≥98.5%
C5H5NOS
MFCD00006196
12,417; 14,268; 17,292
1-Hydroxy-2(1H)-pyridinethione
68° to 72°C

Safety and Handling

Product Identifier
  • 2-Mercaptopyridine-N-oxide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents