missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7081670
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2637-34-5
C5H5NS
111.162
MFCD00006285
WHMDPDGBKYUEMW-UHFFFAOYSA-N
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
2723698
CHEBI:45223
1H-pyridine-2-thione
C1=CC(=S)NC=C1
This item is not returnable. View return policy
2637-34-5
C5H5NS
111.162
MFCD00006285
WHMDPDGBKYUEMW-UHFFFAOYSA-N
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
2723698
CHEBI:45223
1H-pyridine-2-thione
C1=CC(=S)NC=C1

Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.

Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 124°C to 130°C
Quantity 5 g
Beilstein 105758
Sensitivity Air sensitive
Solubility Information Slightly soluble in chloroform,methanol and dimethyl sulfoxide.
Formula Weight 111.17
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'