missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzimidazole, 98%, Thermo Scientific Chemicals

Catalog Number p-155336
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
583-39-1
C7H6N2S
150.20
MFCD00466107
YHMYGUUIMTVXNW-UHFFFAOYSA-N
2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
707035
S=C1NC2=CC=CC=C2N1
This item is not returnable. View return policy
583-39-1
C7H6N2S
150.20
MFCD00466107
YHMYGUUIMTVXNW-UHFFFAOYSA-N
2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
707035
S=C1NC2=CC=CC=C2N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 302°C to 307°C
Color Beige to White
Boiling Point 451°C
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 119
Merck Index 15, 1084
Solubility Information Solubility in water: 2.6g/L (23°C). Other solubilities: soluble in acetone,ethyl acetate and ethanol
IUPAC Name 1,3-dihydrobenzimidazole-2-thione
Formula Weight 150.2
Percent Purity 98%
Insoluble Matter 0.3% max. (in NaOH)
Physical Form Powder
Chemical Name or Material 2-Mercaptobenzimidazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Mercaptobenzimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'