missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-6-nitrobenzothiazole 96%, ACROS Organics™

Brand:  Acros Organics 413610050

Code : 29

Additional Details : CAS Number : 4845-58-3 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10075650

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Mercapto-6-nitrobenzothiazole
4845-58-3
100.0
96%
212.25
96%
95.0
96%
C7H4N2O2S2
MFCD00041850

Specifications

2-Mercapto-6-nitrobenzothiazole
4845-58-3
100.0
Glass bottle
96%
C7H4N2O2S2
MFCD00041850
249.0° to 253.0°C
96%
95.0
Authentic
Powder
96%
212.25
Yellow
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Mercapto-6-nitrobenzothiazole

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS