missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Mercapto-4,6-dimethylpyrimidine hydrate, 98%

Catalog Number p-7081934
Click to view available options
Quantity:
10 g
250 g
This item is not returnable. View return policy
22325-27-5
C6H8N2S
140.204
MFCD00006079
RAFAYWADRVMWFA-UHFFFAOYSA-N
4,6-dimethyl-2-mercaptopyrimidine, 4,6-dimethyl-2 1h-pyrimidinethione, 4,6-dimethylpyrimidine-2-thiol, 4,6-dimethyl-2-thiopyrimidine, 4,6-dimethyl-2-pyrimidinethiol, 2-mercapto-4,6-dimethylpyrimidine, 2 1h-pyrimidinethione, 4,6-dimethyl, 4,6-dimethylpyrimidine-2-thione, 4,6-dimethyl-pyrimidine-2-thiol, 4,6-dimethyl-2-thiolpyrimidine
673664
4,6-dimethyl-1H-pyrimidine-2-thione
CC1=CC(=NC(=S)N1)C
This item is not returnable. View return policy
22325-27-5
C6H8N2S
140.204
MFCD00006079
RAFAYWADRVMWFA-UHFFFAOYSA-N
4,6-dimethyl-2-mercaptopyrimidine, 4,6-dimethyl-2 1h-pyrimidinethione, 4,6-dimethylpyrimidine-2-thiol, 4,6-dimethyl-2-thiopyrimidine, 4,6-dimethyl-2-pyrimidinethiol, 2-mercapto-4,6-dimethylpyrimidine, 2 1h-pyrimidinethione, 4,6-dimethyl, 4,6-dimethylpyrimidine-2-thione, 4,6-dimethyl-pyrimidine-2-thiol, 4,6-dimethyl-2-thiolpyrimidine
673664
4,6-dimethyl-1H-pyrimidine-2-thione
CC1=CC(=NC(=S)N1)C
Melting Point 213°C to 216°C
Quantity 10 g
Beilstein 81330
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'