missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-4,5-diphenyloxazole, 98%, Thermo Scientific™

Catalog Number p-7024867
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
6670-13-9
C15H11NOS
253.319
MFCD00173660
NYQSTRUSYDZNHC-UHFFFAOYSA-N
4,5-diphenyl-2-oxazolethiol, 4,5-diphenyl-1,3-oxazole-2-thiol, diphenyl-1,3-oxazole-2-thiol, 4,5-diphenyl-2-mercaptooxazole, 4,5-diphenyl-1,3-oxazole-2 3h-thione, 4,5-diphenyloxazole-2 3h-thione, 4,5-diphenyl-4-oxazoline-2-thione, 4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione, 4,5-diphenyloxazole-2-thiol, 4,5-diphenyl-3h-oxazole-2-thione
853676
4,5-diphenyl-3H-1,3-oxazole-2-thione
C1=CC=C(C=C1)C2=C(OC(=S)N2)C3=CC=CC=C3
This item is not returnable. View return policy
6670-13-9
C15H11NOS
253.319
MFCD00173660
NYQSTRUSYDZNHC-UHFFFAOYSA-N
4,5-diphenyl-2-oxazolethiol, 4,5-diphenyl-1,3-oxazole-2-thiol, diphenyl-1,3-oxazole-2-thiol, 4,5-diphenyl-2-mercaptooxazole, 4,5-diphenyl-1,3-oxazole-2 3h-thione, 4,5-diphenyloxazole-2 3h-thione, 4,5-diphenyl-4-oxazoline-2-thione, 4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione, 4,5-diphenyloxazole-2-thiol, 4,5-diphenyl-3h-oxazole-2-thione
853676
4,5-diphenyl-3H-1,3-oxazole-2-thione
C1=CC=C(C=C1)C2=C(OC(=S)N2)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 251°C to 256°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 253.33
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4,5-diphenyloxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'