missing translation for 'onlineSavingsMsg'
Learn More

2-Ketoglutaric acid, disodium salt, dihydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-152549
Click to view available options
Quantity:
5 g
50 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
305-72-6
C5H4Na2O5·2H2O
226.1
MFCD00150702
YBGBJYVHJTVUSL-UHFFFAOYSA-L
disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
31040
disodium;2-oxopentanedioate
C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
305-72-6
C5H4Na2O5·2H2O
226.1
MFCD00150702
YBGBJYVHJTVUSL-UHFFFAOYSA-L
disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
31040
disodium;2-oxopentanedioate
C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color White
Loss on Drying 12% to 16%
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NaOOCCH2CH2C(O)COONa·2H2O
Quantity 5 g
Beilstein 03, 1813
Solubility Information Solubility in water: soluble in water
Formula Weight 226.1
Percent Purity 99%
Physical Form Crystalline Powder or Granules
Chemical Name or Material 2-Ketoglutaric acid, disodium salt, dihydrate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'