missing translation for 'onlineSavingsMsg'
Learn More

2-Ketoglutaric acid, 98%, ACROS Organics™

Brand:  Acros Organics 122865000

Code : 29

Additional Details : CAS Number : 328-50-7 Weight : 0.55000kg


Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10666714

  • £124.30 / 500g


Estimated Shipment
07-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Ketoglutaric acid
97.5
≥97.5% (on anhydrous substance)
C5H6O5
146.1
2-Oxoglutaric acid
328-50-7
100.0
98%
HO2CCH2CH2COCO2H
MFCD00004165

Specifications

2-Ketoglutaric acid
97.5
100.0
10ppm max.
30ppm max.
Crystalline Powder
98%
C5H6O5
146.1
03, 789
15, 5350
2-Oxoglutaric acid
95% min. (c=10, H2O)
500g
328-50-7
200ppm max.
200ppm max.
Authentic
1% max. (105°C, 1 hr)
≥97.5% (on anhydrous substance)
Plastic bottle
HO2CCH2CH2COCO2H
MFCD00004165
01,531
0.2% max.
200ppm max.
White to slightly yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Ketoglutaric acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS