missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenol, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081856
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
88-69-7
C9H12O
136.194
MFCD00002224
CRBJBYGJVIBWIY-UHFFFAOYSA-N
2-isopropylphenol, o-hydroxycumene, o-isopropylphenol, o-cumenol, 1-hydroxy-2-isopropylbenzene, 2-propan-2-yl phenol, 2-1-methylethyl phenol, phenol, o-isopropyl, phenol, 2-1-methylethyl, isopropylphenol
6943
CHEBI:38506
2-propan-2-ylphenol
CC(C)C1=CC=CC=C1O
This item is not returnable. View return policy
88-69-7
C9H12O
136.194
MFCD00002224
CRBJBYGJVIBWIY-UHFFFAOYSA-N
2-isopropylphenol, o-hydroxycumene, o-isopropylphenol, o-cumenol, 1-hydroxy-2-isopropylbenzene, 2-propan-2-yl phenol, 2-1-methylethyl phenol, phenol, o-isopropyl, phenol, 2-1-methylethyl, isopropylphenol
6943
CHEBI:38506
2-propan-2-ylphenol
CC(C)C1=CC=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 14°C to 16°C
Density 1.01
Boiling Point 212°C to 214°C
Flash Point 88°C (190°F)
Assay Percent Range 98%
Refractive Index 1.527
Quantity 50 g
UN Number UN2922
Beilstein 1363322
Sensitivity Air sensitive
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 2-Isopropylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'