missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropylaniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Brown or Orange to Yellow |
Density | 0.9550g/mL |
Boiling Point | 112.0°C to 113.0°C (18.0 mmHg) |
Flash Point | 95°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)2CHC6H4NH2 |
Show More |
Safety and Handling
Product Identifier
- 2-Isopropylaniline
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'