missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyphenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 431380100

Code : 29

Additional Details : CAS Number : 138008-97-6 Weight : 0.06000kg


Quantity 10g

 View more versions of this product

Product Code. 11995881

  • £78.50 / 10g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

2-Isopropoxyphenylboronic acid
96.0
97%
C9H13BO3
138008-97-6
100.0
97%
180.01

Specifications

2-Isopropoxyphenylboronic acid
199.0°C
39.0° to 43.0°C
138008-97-6
100.0
Authentic
97%
180.01
110°C
White to off-white
10g
96.0
Crystalline Powder
97%
C9H13BO3
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Isopropoxyphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS