missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyphenol, 97%, Thermo Scientific™

Catalog Number p-151013
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
4812-20-8
MFCD00002184
ZNCUUYCDKVNVJH-UHFFFAOYSA-N
2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a
20949
CHEBI:38547
2-propan-2-yloxyphenol
CC(C)OC1=CC=CC=C1O
This item is not returnable. View return policy
4812-20-8
MFCD00002184
ZNCUUYCDKVNVJH-UHFFFAOYSA-N
2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a
20949
CHEBI:38547
2-propan-2-yloxyphenol
CC(C)OC1=CC=CC=C1O
CAS Min % 96.0
CAS Max % 100.0
Color Brown-Red to Yellow
Density 1.0300g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C9H12O2
Linear Formula (CH3)2CHOC6H4OH
Refractive Index 1.5140 to 1.5160 (20°C, 589nm)
Quantity 5 g
Molecular Weight (g/mol) 152.193
Formula Weight 152.19
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2-Isopropoxyphenol, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'