missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Isobutylpyrazine, 98%

Brand:  Alfa Aesar™ A10245.03

Code : NEW

Additional Details : CAS Number : 29460-92-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11403953

  • £24.10 / 1g


Estimated Shipment date
from Supplier 22-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Isobutylpyrazine
C8H12N2
MFCD00156150
29460-92-2
136.2

Specifications

2-Isobutylpyrazine
29460-92-2
C8H12N2
MFCD00156150
1g
98%
136.2
Videos
SDS