missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Isobutylpyrazine, 98%

Brand:  Alfa Aesar™ A10245.14

Code : NEW

Additional Details : CAS Number : 29460-92-2 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11423953

  • £311.10 / 25g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Isobutylpyrazine
C8H12N2
MFCD00156150
29460-92-2
136.2

Specifications

2-Isobutylpyrazine
29460-92-2
C8H12N2
MFCD00156150
25g
98%
136.2
Videos
SDS