missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoxybenzoic acid, stabilized, Thermo Scientific Chemicals

Catalog Number p-3211717
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
61717-82-6, 64297-64-9, 121-91-5, 65-85-0
C7H5IO4
280.01
MFCD00024166
61717-82-6, 64297-64-9, 121-91-5, 65-85-0
C7H5IO4
280.01
MFCD00024166

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 250.0°C
Color White
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 5091
Solubility Information Solubility in water: slightly soluble
Formula Weight 280.01
Percent Purity 39 to 50% as 2-Iodoxybenzoic acid
Physical Form Powder, Prills or Agglomerates
Chemical Name or Material 2-Iodoxybenzoic acid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Iodoxybenzoic acid, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'