missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Iodopyridine, 95%

Brand:  Alfa Aesar™ H63062.06

Code : NEW

Additional Details : CAS Number : 5029-67-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449538

  • £40.50 / 5g


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Description

Description

Applications
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2-Iodopyridine
205
105790
5029-67-4
MFCD00464928

Specifications

2-Iodopyridine
1.632
95%
205
105790
52°C (dec.)
Light Sensitive
5029-67-4
Solid
1.928g/mL
MFCD00464928
Pale yellow
5g
Videos
SDS