missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
29.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
2-Iodophenyl isothiocyanate is an important raw material and intermediate used in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodophenyl isothiocyanate is an important raw material and intermediate used in organic synthesis and pharmaceuticals.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light and moisture. Incompatible with polyethers, polyester, alcohols, bases, amines and strong oxidizing agents.
2-Iodophenyl isothiocyanate is an important raw material and intermediate used in organic synthesis and pharmaceuticals.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light and moisture. Incompatible with polyethers, polyester, alcohols, bases, amines and strong oxidizing agents.
Specifications
Melting Point | 36°C to 39°C |
Flash Point | >110°C (230°F) |
Quantity | 1 g |
UN Number | UN2923 |
Beilstein | 2716369 |
Sensitivity | Light and moisture sensitive |
Formula Weight | 261.09 |
Percent Purity | 97% |
Chemical Name or Material | 2-Iodophenyl isothiocyanate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'