missing translation for 'onlineSavingsMsg'
Learn More

2-Iodomelatonin, Tocris Bioscience™

High affinity melatonin agonist

Brand:  Tocris (Bio-Techne) 073750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437933

  • £264.60 / 50mg

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent melatonin agonist (pKi values are 10.55 and 9.87 at human recombinant MT1 and MT2 receptors respectively). 2-[125I]-iodomelatonin has been used to identify, characterize and localize melatonin binding sites in the brain and peripheral tissues.
Specifications

Chemical Identifiers

93515-00-5
358.179
2-iodomelatonin, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl acetamide, 2-iodo-melatonin, 2-i-mlt, acetamide, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl-, 3beta, acetamide,n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl, n-2-2-iodo-5-methoxyindol-3-yl ethyl acetamide, acetamide, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl, melatonin,2-iodo
N-[2-(2-iodo-5-methoxy-1H-indol-3-yl)ethyl]acetamide
C13H15IN2O2
FJDDSMSDZHURBJ-UHFFFAOYSA-N
115348
CC(=O)NCCC1=C(NC2=C1C=C(C=C2)OC)I

Specifications

50mg
93515-00-5
2-iodomelatonin, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl acetamide, 2-iodo-melatonin, 2-i-mlt, acetamide, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl-, 3beta, acetamide,n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl, n-2-2-iodo-5-methoxyindol-3-yl ethyl acetamide, acetamide, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl, melatonin,2-iodo
CC(=O)NCCC1=C(NC2=C1C=C(C=C2)OC)I
358.179
358.18
2-Iodomelatonin
C13H15IN2O2
FJDDSMSDZHURBJ-UHFFFAOYSA-N
N-[2-(2-iodo-5-methoxy-1H-indol-3-yl)ethyl]acetamide
115348
>98%
Videos
Documents